missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGLUT1 Antibody (SR2075), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21883-50ul
This item is not returnable.
View return policy
Description
VGLUT1 Monoclonal antibody specifically detects VGLUT1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| VGLUT1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR2075 | |
| Western Blot 1:500-1:2000 | |
| BNPIbrain-specific Na-dependent inorganic phosphate cotransporter, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, member 7, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), Solute carrier family 17 member 7, VGluT1, VGLUT1vesicular glutamate transporter 1 | |
| A synthesized peptide derived from human VGLUT1 (Uniprot #: Q9P2U7) | |
| 50 μg | |
| ABC Transporters, Cellular Signaling, Neuronal Cell Markers, Neuroscience | |
| 57030 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion