missing translation for 'onlineSavingsMsg'
Learn More

VGLUT1 Antibody (SR2075), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 338.00 - € 449.00

Specifications

Antigen VGLUT1
Clone SR2075
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18683055
View Documents
Novus Biologicals
NBP3-21883-100ul
100 μg
€ 449.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18673744
View Documents
Novus Biologicals
NBP3-21883-50ul
50 μg
€ 338.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

VGLUT1 Monoclonal antibody specifically detects VGLUT1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

VGLUT1
Western Blot 1:500-1:2000
Monoclonal
Purified
RUO
Human, Mouse, Rat
BNPIbrain-specific Na-dependent inorganic phosphate cotransporter, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, member 7, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), Solute carrier family 17 member 7, VGluT1, VGLUT1vesicular glutamate transporter 1
A synthesized peptide derived from human VGLUT1 (Uniprot #: Q9P2U7)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR2075
Western Blot
Unconjugated
Rabbit
ABC Transporters, Cellular Signaling, Neuronal Cell Markers, Neuroscience
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
57030
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.