Learn More
Applied Biosystems™ ViiA™ 7 Dx Block Upgrades
Front access makes it easy to change thermal cycling block formats without having to move attached peripherals such as robots or computers.
Brand: Applied Biosystems™ 4453550
Includes: Includes 384-well Dx Block, heated cover, plate adaptor and calibration kit.
Description
The ViiA™ 7 Dx Instrument puts high-productivity qPCR at your fingertips. With the new OptiFlex™ optical system and 4 interchangeable block formats from 96-well to 384-well to Micro Fluidic Array Card, the ViiA™ 7 Dx Instrument is available for in-vitro diagnostics use.
Reaction Speed: Standard