Learn More
Applied Biosystems™ ViiA™ 7 Dx Block Upgrades
Front access makes it easy to change thermal cycling block formats without having to move attached peripherals such as robots or computers.
€ 18820.00 - € 33470.00
Includes: Includes 384-well Dx Block, heated cover, plate adaptor and calibration kit.
| Product Code | Brand | Quantity | Description | Includes | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Description | Includes | Price | Quantity & Availability | |||
|
15873304
|
Applied Biosystems™
4453549 |
€ 18820.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
15949222
|
Applied Biosystems™
4453550 |
€ 33470.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
The ViiA™ 7 Dx Instrument puts high-productivity qPCR at your fingertips. With the new OptiFlex™ optical system and 4 interchangeable block formats from 96-well to 384-well to Micro Fluidic Array Card, the ViiA™ 7 Dx Instrument is available for in-vitro diagnostics use.
Reaction Speed: StandardYour input is important to us. Please complete this form to provide feedback related to the content on this product.