Learn More
Applied Biosystems™ ViiA™ 7 Dx Block Upgrades
Front access makes it easy to change thermal cycling block formats without having to move attached peripherals such as robots or computers.
€ 18820.00 - € 33470.00
Includes: Includes 384-well Dx Block, heated cover, plate adaptor and calibration kit.
Products 2
Product Code Brand Quantity Description Includes Price Quantity & Availability  
Product Code Brand Quantity Description Includes Price Quantity & Availability  
15873304
View Documents
Applied Biosystems™
4453549
€ 18820.00
Each
View alternative products
Save Up to 
»
15949222
View Documents
Applied Biosystems™
4453550
€ 33470.00
Each
View alternative products
Save Up to 
»
Description
Description
The ViiA™ 7 Dx Instrument puts high-productivity qPCR at your fingertips. With the new OptiFlex™ optical system and 4 interchangeable block formats from 96-well to 384-well to Micro Fluidic Array Card, the ViiA™ 7 Dx Instrument is available for in-vitro diagnostics use.
Reaction Speed: Standard