missing translation for 'onlineSavingsMsg'
Learn More

Vinblastine sulfate, MedChemExpress

Product Code. 16432007
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16432007 50 mg 50mg
16488507 5 mg 5mg
16456326 100 mg 100mg
16425858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16432007 Supplier Medchem Express Supplier No. HY13780

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vinblastine sulfate is a cytotoxic alkaloid used against various cancer types. Vinblastine sulfate inhibits the formation of microtubule and suppresses nAChR with an IC50 of 8.9 μM.

Vinblastine sulfate is a cytotoxic alkaloid used against various cancer types. Vinblastine sulfate inhibits the formation of microtubule and suppresses nAChR with an IC50 of 8.9 μM.

Specifications

Chemical Name or Material Vinblastine sulfate
CAS 143-67-9
Color White
Purity Grade Notes Research
Molecular Formula C46H60N4O13S
Quantity 50 mg
Synonym Vincaleukoblastine sulfate salt
Solubility Information H2O : 50 mg/mL (55.00 mM; Need ultrasonic) ∣DMSO : ≥ 44 mg/mL (48.40 mM)
SMILES [H][C@@]12C[C@@](C3=C(OC)C=C(N([C@]4([H])[C@@]56[C@]7([H])[C@](C=CCN7CC6)(CC)[C@@H](OC(C)=O)[C@@]4(C(OC)=O)O)C)C5=C3)(C(OC)=O)C(NC8=C9C=CC=C8)=C9CC[N@](C2)C[C@](O)(CC)C1.OS(=O)(O)=O
Molecular Weight (g/mol) 909.05
Formula Weight 909.05
Percent Purity 98.86%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.