Learn More
Ansell™ Virtex™ 79-700 Series Blue Nitrile Immersion Gloves
Product Code. 15262709 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
200 pairs
50 pairs
Product Code. 15262709
Brand: Ansell™ 79700_10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed specifically to offer outstanding comfort, flexibility and good chemical resistance. Ansell Virtex™ 79-700 Series Blue Nitrile Immersion Gloves provide all-day comfort in a lightweight glove.
  • Reusable
  • Anti-static according to EN1149
  • With Aquadri™-Ansell Moisture Management Technology™, which gives excellent moisture absorption capabilities and keeps users' hands drier for longer
  • Significantly lower tendency to shed than traditional cotton flock gloves
  • Ideal for light assembly in dry, fully wet or oily conditions, engine and system testing, paint and spray shops, food distribution
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Cuff Material Nitrile
Product Type Immersion Gloves
Size 10
Style 79-700
Thickness (Metric) 0.2 mm
Glove Cuff Color Blue
Glove Cuff Style Straight
Liner Material Aquadri
Quantity 50 Pairs
Resistant To Good chemical resistance
Series 79-700