missing translation for 'onlineSavingsMsg'
Learn More

Vitamin D binding protein Mouse anti-Human, Clone: 1E4D10, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66175-1-IG-20UL

 View more versions of this product

Product Code. 16807815

  • € 145.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.
Specifications

Specifications

Vitamin D binding protein
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02774
GC
Vitamin D binding protein Fusion Protein Ag9803
20 μL
Primary
Human
Antibody
IgG1
Western Blot
1E4D10
Unconjugated
GC
DBP, DBP/GC, GC, Gc globulin, VDBP,GC, Vitamin D binding protein
Mouse
Protein G
RUO
2638
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.