missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitamin D binding protein Mouse anti-Human, Clone: 1E4D10, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | Vitamin D binding protein |
|---|---|
| Clone | 1E4D10 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16807815
|
Proteintech
66175-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16897805
|
Proteintech
66175-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.Specifications
| Vitamin D binding protein | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| DBP, DBP/GC, GC, Gc globulin, VDBP,GC, Vitamin D binding protein | |
| GC | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1E4D10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P02774 | |
| 2638 | |
| Vitamin D binding protein Fusion Protein Ag9803 | |
| Primary | |
| -20°C | |
| GC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title