missing translation for 'onlineSavingsMsg'
Learn More

Vitamin D4, MedChemExpress

Product Code. 16441969
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16441969

Brand: Medchem Express HY75958

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vitamin D4 (22-Dihydroergocalciferol) is a Vitamin D derived from fungi. The precursor of Vitamin D4 is 22,23-dihydroergosterol.

Vitamin D4 (22-Dihydroergocalciferol) is a Vitamin D derived from fungi. The precursor of Vitamin D4 is 22,23-dihydroergosterol.

Specifications

Chemical Name or Material Vitamin D4
CAS 511-28-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H46O
Quantity 1 mg
Synonym 22-Dihydroergocalciferol
Solubility Information DMSO : 25 mg/mL (62.71 mM; Need ultrasonic)
SMILES C=C1/C(C[C@@H](O)CC1)=C\C=C2[C@@]3([H])[C@@](CCC\2)(C)[C@@H]([C@H](C)CC[C@H](C)C(C)C)CC3
Molecular Weight (g/mol) 398.66
Formula Weight 398.66
Percent Purity 97.98%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.