Learn More
Poulten Graf™ Volac™ Class A Borosilicate Glass Burettes with Certificate and PTFE Key
Brand:  Poulten Graf™ RB9806/WAC/Y
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 11877990
  • € 79.60 / Each
Estimated Shipment: 02-05-2024
to see stock.
Specifications
10 mL
0.02 mL
Blue
PTFE
Class AS
Borosilicate Glass
Interchangeable PTFE Key in Glass Barrel
Straight
Volumetric Buret
BS 846, ISO 385