Learn More
Poulten Graf™ Volac™ Class A Borosilicate Glass Burettes with Certificate and PTFE Key
Brand: Poulten Graf™ RB9806/WAC/I
Additional Details : Weight : 0.80000kg
Specifications
100 mL | |
0.2 mL | |
Blue | |
PTFE | |
Class AS |
Borosilicate Glass | |
Interchangeable PTFE Key in Glass Barrel | |
Straight | |
Volumetric Buret | |
BS 846, ISO 385 |