Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Blue Graduation
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Blue Graduation is ideal for measuring and transferring various quantities of liquid.
Brand:  Poulten & Graf GmbH 1 11427402
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10381602
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Blue Graduation is ideal for measuring and transferring various quantities of liquid.
  • Total delivery
  • Blue graduation
  • With ring marks
  • Comformity approved
Specifications
Specifications
EN ISO 835
Graduated pipette
Non-sterile
Blue
2 mL
Reusable
AS
Soda Lime Glass
Yes
0.01 mL
Graduated Pipet