missing translation for 'onlineSavingsMsg'
Learn More

Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Blue Graduation

Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Blue Graduation is ideal for measuring and transferring various quantities of liquid.

Brand:  Poulten & Graf GmbH 1 11433402

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15251129

  • € 16.95 / Pack of 5
Estimated Shipment: 23-05-2024
to see stock.
Add to basket

Description

Description

Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Blue Graduation is ideal for measuring and transferring various quantities of liquid.

  • Total delivery
  • Blue graduation
  • With ring marks
  • Comformity approved
Specifications

Specifications

EN ISO 835
Graduated pipette
Non-sterile
Blue
5 mL
Reusable
AS
Soda Lime Glass
Yes
0.03 mL
Graduated Pipet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers