Learn More
Poulten & Graf GmbH VOLAC USP Graduated Cylinder
Safely and accurately aquire measurements for all laboratory needs. Poulten & Graf GmbH VOLAC USP Graduated Cylinder measures the volume of a liquid using horizontal marked lines.
Brand:  Poulten & Graf GmbH US263/WAC/B/2
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10486562
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Hexagonal base
  • White graduation
  • With individual serial number
  • With single certificate
Specifications
Specifications
Round
Graduated Cylinder
Blue
Hexagonal
Volac Graduated Cylinder, USP, with single certificate
10 mL
0.2 mL
10mL
0.2mL
±0.08 mL
To Contain
EN ISO 4788
A
Borosilicate Glass
White
Autoclavable
0.08mL