missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WASL Monoclonal Antibody (OTI9F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811284S
This item is not returnable.
View return policy
Description
WASL Monoclonal antibody specifically detects WASL in Human samples. It is validated for Western BlotSpecifications
| WASL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00401 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8976 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI9F2 | |
| Unconjugated | |
| WASL | |
| 2900021I12Rik; 3110031I02Rik; DKFZp779G0847; MGC48327; n wasp; neural Wiskott-Aldrich syndrome protein; NWASP; N-WASP; TRS4; WASL; WASP like actin nucleation promoting factor; WASPB; Wiskott-Aldrich syndrome like; Wiskott-Aldrich syndrome-like; Wiskott-Aldrich syndrome-like (human) | |
| Full length human recombinant protein of human WASL produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction