missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WASL Monoclonal Antibody (OTI9F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811284
This item is not returnable.
View return policy
Description
The Wiskott-Aldrich syndrome (WAS) family of proteins share similar domain structure, and are involved in transduction of signals from receptors on the cell surface to the actin cytoskeleton. The presence of a number of different motifs suggests that they are regulated by a number of different stimuli, and interact with multiple proteins. Recent studies have demonstrated that these proteins, directly or indirectly, associate with the small GTPase, Cdc42, known to regulate formation of actin filaments, and the cytoskeletal organizing complex, Arp2/3. The WASL gene product is a homolog of WAS protein, however, unlike the latter, it is ubiquitously expressed and shows highest expression in neural tissues. It has been shown to bind Cdc42 directly, and induce formation of long actin microspikes.Specifications
| WASL | |
| Monoclonal | |
| Unconjugated | |
| WASL | |
| N-WASP, NWASP, WASPB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 73178, 8976 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI9F2 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00401, Q91YD9 | |
| WASL | |
| Full length human recombit protein of human WASL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction