Learn More
Heathrow Scientific Weighing Papers, White
Shop All Fisherbrand Products
Click to view available options
Length (Metric):
10.2 cm
15.2 cm
Unit Size:
Pack of 500
Specifications
Specifications
| Description | Weighing Papers |
| Length (English) | 4 in. |
| Length (Metric) | 10.2 cm |
| Material | Paper |
| Width (English) | 4 in. |
| Width (Metric) | 10.2 cm |
| Product Type | Weighing Paper |
| Shape | Square |