Learn More
DWK Life Sciences Wheaton™ Unlined Aluminum Open-Top Seals
GREENER_CHOICE
Product Code. p-77096 Shop All Wheaton Science Products Products
Click to view available options
Color:
Blue
Green
Natural
Red
Closure Size:
11 mm
13 mm
20 mm
Unit Size:
Pack of 1000
Product Code. 11788769
Brand: DWK Life Sciences 22417805
Only null left
Add to basket
Add to basket
Open-top seal exposes stopper for sample retrieval with a syringe
  • Unlined
  • Closure Type: Seal
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
Material Aluminum
Quantity 1000/Cs.
Type Open Top Cap
Closure Size 20 mm