missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIF-1 Antibody (SR1697), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22234-100ul
This item is not returnable.
View return policy
Description
WIF-1 Monoclonal antibody specifically detects WIF-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| WIF-1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1697 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| WIF-1, WNT inhibitory factor 1 | |
| A synthesized peptide derived from human WIF-1 (Uniprot #: Q9Y5W5 ) | |
| 100 μg | |
| Signal Transduction, Wnt Signaling Pathway | |
| 11197 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction