missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIF-1 Antibody (SR1697), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | WIF-1 |
|---|---|
| Clone | SR1697 |
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18614275
|
Novus Biologicals
NBP3-22234-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18641894
|
Novus Biologicals
NBP3-22234-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WIF-1 Monoclonal antibody specifically detects WIF-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| WIF-1 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| WIF-1, WNT inhibitory factor 1 | |
| A synthesized peptide derived from human WIF-1 (Uniprot #: Q9Y5W5 ) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1697 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction, Wnt Signaling Pathway | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 11197 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title