missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wilm s Tumor 1 Mouse, Clone: rWT1/857, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 7490-MSM6-P0
This item is not returnable.
View return policy
Specifications
| Wilm s Tumor 1 | |
| Cytoplasmic | |
| rWT1/857 | |
| Unconjugated | |
| NP_001185480 | |
| Mouse | |
| 47-55kDa | |
| Protein A/G Purified | |
| RUO | |
| 7490 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| WT1 | |
| Recombinant full-length human WT1 protein | |
| K562 cells. Wilm s Tumor, mesothelioma or fetal kidney. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction