missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wilm s Tumor 1 Mouse, Clone: rWT1/857, enQuireBio™
Mouse Monoclonal Antibody
€ 199.00 - € 478.00
Specifications
| Antigen | Wilm s Tumor 1 |
|---|---|
| Clone | rWT1/857 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15981088
|
EnquireBio
7490-MSM6-P0 |
20 μg | |||||||
|
15991088
|
EnquireBio
7490-MSM6-P1 |
100 μg |
€ 573.00 € 478.00 / 100µg Save € 95.00 16% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| Wilm s Tumor 1 | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| 7490 | |
| Recombinant full-length human WT1 protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 47-55kDa |
| rWT1/857 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_001185480 | |
| WT1 | |
| IgG1 κ | |
| Protein A/G Purified | |
| Cytoplasmic | |
| K562 cells. Wilm s Tumor, mesothelioma or fetal kidney. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title