missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIPI1 Antibody (SR1849), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21991-100ul
This item is not returnable.
View return policy
Description
WIPI1 Monoclonal antibody specifically detects WIPI1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| WIPI1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1849 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100 | |
| Atg18, Atg18 protein homolog, ATG18A, FLJ10055, WD repeat domain phosphoinositide-interacting protein 1, WD repeat domain, phosphoinositide interacting 1, WD40 repeat protein interacting with phosphoinositides of 49 kDa, WD40 repeat protein Interacting with phosphoInositides of 49kDa, WIPI 49 kDa, WIPI-1, WIPI-1 alpha, WIPI49ATG18 | |
| A synthesized peptide derived from human WIPI1 (Uniprot #: Q5MNZ9) | |
| 100 μg | |
| Autophagy, Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| 55062 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction