missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIPI1 Antibody (SR1849), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | WIPI1 |
|---|---|
| Clone | SR1849 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18664355
|
Novus Biologicals
NBP3-21991-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18602105
|
Novus Biologicals
NBP3-21991-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WIPI1 Monoclonal antibody specifically detects WIPI1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| WIPI1 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Atg18, Atg18 protein homolog, ATG18A, FLJ10055, WD repeat domain phosphoinositide-interacting protein 1, WD repeat domain, phosphoinositide interacting 1, WD40 repeat protein interacting with phosphoinositides of 49 kDa, WD40 repeat protein Interacting with phosphoInositides of 49kDa, WIPI 49 kDa, WIPI-1, WIPI-1 alpha, WIPI49ATG18 | |
| A synthesized peptide derived from human WIPI1 (Uniprot #: Q5MNZ9) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1849 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 55062 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title