missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YY1AP1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94366-0.1ml
This item is not returnable.
View return policy
Description
YY1AP1 Polyclonal antibody specifically detects YY1AP1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| YY1AP1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ10875, FLJ13914, HCCA1, HCCA2YY1APYAP, Hepatocellular carcinoma susceptibility protein, Hepatocellular carcinoma-associated protein 2, YY1 associated protein 1, YY1-associated protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human YY1AP1 (NP_060723.2). MGFSNMEDDGPEEEERVAEPQANFNTPQALRFEELLANLLNEQHQIAKELFEQLKMKKPSAKQQKEVEKVKPQCKEVHQT | |
| 0.1 mL | |
| Cancer | |
| 55249 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction