missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YY1AP1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 470.00
Specifications
| Antigen | YY1AP1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18696842
|
Novus Biologicals
NBP2-94366-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18639441
|
Novus Biologicals
NBP2-94366-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
YY1AP1 Polyclonal antibody specifically detects YY1AP1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| YY1AP1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS (pH 7.3), 50% glycerol | |
| 55249 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ10875, FLJ13914, HCCA1, HCCA2YY1APYAP, Hepatocellular carcinoma susceptibility protein, Hepatocellular carcinoma-associated protein 2, YY1 associated protein 1, YY1-associated protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human YY1AP1 (NP_060723.2). MGFSNMEDDGPEEEERVAEPQANFNTPQALRFEELLANLLNEQHQIAKELFEQLKMKKPSAKQQKEVEKVKPQCKEVHQT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title