Learn More
[(Z)-Prop-1-enyl]phosphonic Acid (Technical Grade), TRC
Product Code. 30450621
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450621 5 g 5g
30455521 1 g 1g
30451824 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30450621 Supplier Toronto Research Chemicals Supplier No. TRCZ1927005G
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25383-06-6
Molecular Formula C3H7O3P
Molecular Weight (g/mol) 122.06
Synonym Phosphonic acid, P-(1Z)-1-propen-1-yl- (ACI), P-(1Z)-1-Propen-1-ylphosphonic acid (ACI), Phosphonic acid, (1Z)-1-propenyl- (9CI), Phosphonic acid, 1-propenyl-, (Z)- (ZCI), Phosphonic acid, propenyl-, (Z)- (8CI), (cis-1-Propenyl)phosphonic acid, [(Z)-1-Propenyl]phosphonic acid, cis-Propenylphosphonic acid
IUPAC Name [(Z)-prop-1-enyl]phosphonic acid
SMILES C\C=C/P(=O)(O)O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C3H7O3P/c1-2-3-7(4,5)6/h2-3H,1H3,(H2,4,5,6)/b3-2-
Formula Weight 122.0133
Chemical Name or Material [(Z)-Prop-1-enyl]phosphonic Acid (Technical Grade)