missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZADH1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38122
This item is not returnable.
View return policy
Description
ZADH1 Polyclonal specifically detects ZADH1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ZADH1 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q8N8N7 | |
| PTGR2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VAGKIGHFLGCSRVVGICGTHEKCILLTSELGFDAAINYKKDNVAEQLRESCPAGVDVYFDNVGGNISDTVISQMNENSHIILCGQISQYNKDVPYPPPLSPAIEAIQKERNITRERFLVLNYKDKFEPGILQLSQWFKE | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DKFZp686P10120, EC 1.3.1.48, FLJ39091, FLJ99229,15-oxoprostaglandin 13-reductase, PGR2, PRG-2, prostaglandin reductase 2, ZADH115-oxoprostaglandin-delta13-reductase, zinc binding alcohol dehydrogenase domain containing 1, zinc binding alcohol dehydrogenase, domain containing 1, Zinc-binding alcohol dehydrogenase domain-containing protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 145482 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction