missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZADH1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 572.00
Specifications
| Antigen | ZADH1 |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18652776
|
Novus Biologicals
NBP2-38122-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18188288
|
Novus Biologicals
NBP2-38122 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZADH1 Polyclonal specifically detects ZADH1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ZADH1 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DKFZp686P10120, EC 1.3.1.48, FLJ39091, FLJ99229,15-oxoprostaglandin 13-reductase, PGR2, PRG-2, prostaglandin reductase 2, ZADH115-oxoprostaglandin-delta13-reductase, zinc binding alcohol dehydrogenase domain containing 1, zinc binding alcohol dehydrogenase, domain containing 1, Zinc-binding alcohol dehydrogenase domain-containing protein 1 | |
| PTGR2 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8N8N7 | |
| 145482 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VAGKIGHFLGCSRVVGICGTHEKCILLTSELGFDAAINYKKDNVAEQLRESCPAGVDVYFDNVGGNISDTVISQMNENSHIILCGQISQYNKDVPYPPPLSPAIEAIQKERNITRERFLVLNYKDKFEPGILQLSQWFKE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title