missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZAG Antibody (SR1742), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22121-100ul
This item is not returnable.
View return policy
Description
ZAG Monoclonal antibody specifically detects ZAG in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| ZAG | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| SR1742 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:40 | |
| alpha-2-glycoprotein 1, zinc, alpha-2-glycoprotein 1, zinc-binding, ZA2G, ZAGzn-alpha-2-GP, zinc, zinc-alpha-2-glycoprotein, zn-alpha-2-glycoprotein, ZNGP1 | |
| A synthesized peptide derived from human ZAG (Uniprot #: P25311) | |
| 100 μg | |
| Endocrinology, Immunology, Innate Immunity, Signal Transduction | |
| 563 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction