missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZAG Antibody (SR1742), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | ZAG |
|---|---|
| Clone | SR1742 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:40 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18683594
|
Novus Biologicals
NBP3-22121-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640874
|
Novus Biologicals
NBP3-22121-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZAG Monoclonal antibody specifically detects ZAG in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunoprecipitationSpecifications
| ZAG | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:40 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| alpha-2-glycoprotein 1, zinc, alpha-2-glycoprotein 1, zinc-binding, ZA2G, ZAGzn-alpha-2-GP, zinc, zinc-alpha-2-glycoprotein, zn-alpha-2-glycoprotein, ZNGP1 | |
| A synthesized peptide derived from human ZAG (Uniprot #: P25311) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1742 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Immunology, Innate Immunity, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 563 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title