Learn More
ZFP36L1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1H6G6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-68038-20UL
 View more versions of this product
Product Code. 18018735
  • € 140.00 / 20µL
Description
The ZFP36L1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZFP36L1. This antibody recognizes human, rat antigen. The ZFP36L1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
ZFP36L1
1H6G6
CoraLite Plus 488
Mouse
Primary
Human, Rat
155 kDa
IgG2b
Immunofluorescence, Flow Cytometry (Intracellular Staining)
1000 μg/mL
Berg36, BRF1, Butyrate response factor 1, cMG1, EGF response factor 1, ERF 1, ERF1, Protein TIS11B, RNF162B, TIS11B, ZFP36L1
20 μL
677
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.