missing translation for 'onlineSavingsMsg'
Learn More

ZFP36L1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1H6G6, Proteintech

Mouse Monoclonal Antibody

€ 145.00 - € 450.00

Specifications

Antigen ZFP36L1
Clone 1H6G6
Concentration 1000 μg/mL
Applications Immunofluorescence, Flow Cytometry (Intracellular Staining)
Conjugate CoraLite Plus 488
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18018735
View Documents
Proteintech
CL488-68038-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18045105
View Documents
Proteintech
CL488-68038-100UL
100 μL
€ 450.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The ZFP36L1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZFP36L1. This antibody recognizes human, rat antigen. The ZFP36L1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.

Specifications

Specifications

ZFP36L1
1000 μg/mL
CoraLite Plus 488
Mouse
677
IgG2b
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
1H6G6
Immunofluorescence, Flow Cytometry (Intracellular Staining)
Liquid
Human, Rat
Berg36, BRF1, Butyrate response factor 1, cMG1, EGF response factor 1, ERF 1, ERF1, Protein TIS11B, RNF162B, TIS11B, ZFP36L1
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.