missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zic2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93517-0.1ml
This item is not returnable.
View return policy
Description
Zic2 Polyclonal antibody specifically detects Zic2 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| Zic2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| HPE5Zic family member 2 (odd-paired Drosophila homolog), Zic family member 2 (odd-paired homolog, Drosophila), Zinc finger protein of the cerebellum 2, zinc finger protein ZIC 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ZIC2 (NP_009060.2). MLLDAGPQFPAIGVGSFARHHHHSAAAAAAAAAEMQDRELSLAAAQNGFVDSAAAHMGAFKLNPGAHELSPGQSSAFTSQGPGAYPGSAAAAAAAAALGPHAAHVGSYSGPPFNSTRDFLFRSRGFGDSAPGGGQHGLFGPGAGGLHHAHSDAQGHLLFPGLPEQHGPHGSQNVLNGQMRLGLPGEVFGRSEQYRQVASPRTDPYSAAQLHNQYGPMNMN | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| 7546 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction