missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zic2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 468.00
Specifications
| Antigen | Zic2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Zic2 Polyclonal antibody specifically detects Zic2 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| Zic2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| PBS (pH 7.3), 50% glycerol | |
| 7546 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| HPE5Zic family member 2 (odd-paired Drosophila homolog), Zic family member 2 (odd-paired homolog, Drosophila), Zinc finger protein of the cerebellum 2, zinc finger protein ZIC 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human ZIC2 (NP_009060.2). MLLDAGPQFPAIGVGSFARHHHHSAAAAAAAAAEMQDRELSLAAAQNGFVDSAAAHMGAFKLNPGAHELSPGQSSAFTSQGPGAYPGSAAAAAAAAALGPHAAHVGSYSGPPFNSTRDFLFRSRGFGDSAPGGGQHGLFGPGAGGLHHAHSDAQGHLLFPGLPEQHGPHGSQNVLNGQMRLGLPGEVFGRSEQYRQVASPRTDPYSAAQLHNQYGPMNMN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title