missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZIC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21717-1-AP-150UL
This item is not returnable.
View return policy
Description
ZIC3 is a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in the gene encoding ZIC3 cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs.Specifications
| ZIC3 | |
| Polyclonal | |
| Unconjugated | |
| ZIC3 | |
| HTX, HTX1, ZIC3, Zinc finger protein ZIC 3, ZNF203 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22773, 7547 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60481, Q62521 | |
| ZIC3 | |
| ZIC3 Fusion Protein Ag16354 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction