missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZIC3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | ZIC3 |
|---|---|
| Concentration | 0.24 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16831315
|
Proteintech
21717-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16821315
|
Proteintech
21717-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZIC3 is a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in the gene encoding ZIC3 cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs.Specifications
| ZIC3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| O60481, Q62521 | |
| 22773, 7547 | |
| ZIC3 Fusion Protein Ag16354 | |
| Primary | |
| -20°C | |
| ZIC3 |
| 0.24 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| HTX, HTX1, ZIC3, Zinc finger protein ZIC 3, ZNF203 | |
| ZIC3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title