missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate Dihydrate, Reagent Grade, Honeywell™

Product Code. p-7120908 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 15618210

Brand: Honeywell Research Chemicals Z0625500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5970-45-6
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.498
MDL Number MFCD00066961
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

Melting Point ∼100°C
Color Gray
pH 6.0 to 8.0 (at 20°C)
Quantity 500 g
Linear Formula Zn(CH3COO)2 · 2H2O
UN Number UN3077
Beilstein 3732513
Formula Weight 219.51g/mol
Odor Vinegar-like
Packaging Poly Bottle
Appearance Crystalline
Density 1.750g/cu.cm at 20°C
Chemical Name or Material Zinc acetate dihydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.