missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc finger protein 774 Antibody (PCRP-ZNF774-3F7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20472-100ug
This item is not returnable.
View return policy
Description
Zinc finger protein 774 Monoclonal antibody specifically detects Zinc finger protein 774 in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| Zinc finger protein 774 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Flow Cytometry 0.5-2 μg/million cells, ELISA | |
| MGC75360, zinc finger protein 774 | |
| Recombinant full-length human Zinc finger protein 774 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| PCRP-ZNF774-3F7 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 342132 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction