missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc finger protein 774 Antibody (PCRP-ZNF774-3F7), Novus Biologicals™
Mouse Monoclonal Antibody
€ 249.00 - € 494.00
Specifications
| Antigen | Zinc finger protein 774 |
|---|---|
| Clone | PCRP-ZNF774-3F7 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 0.5-2 μg/million cells, ELISA |
| Applications | Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18639454
|
Novus Biologicals
NBP3-20472-100ug |
100 μg |
€ 494.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18626994
|
Novus Biologicals
NBP3-20472-20ug |
20 μg |
€ 249.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zinc finger protein 774 Monoclonal antibody specifically detects Zinc finger protein 774 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| Zinc finger protein 774 | |
| 0.2 mg/ml | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| MGC75360, zinc finger protein 774 | |
| Recombinant full-length human Zinc finger protein 774 protein | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
| PCRP-ZNF774-3F7 | |
| Flow Cytometry 0.5-2 μg/million cells, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM PBS with 0.05% BSA | |
| 342132 | |
| IgG | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title