missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, RoHS-Monitor, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Zinc, RoHS-Monitor, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Lead/Mercury/Cadmium/Chromium |
| CAS | 7440-66-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Lead 0.0077 %/Mercury 0.029 %/Cadmium 0.215 %/Chromium <0.0002 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction