Learn More
Zinc Sulfate Solution 0.05M (0.05N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 Ampule
6 Ampules
Unit Size:
1 ampoule
6 ampoules
Specifications
Specifications
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Quantity | 1 Ampule |
| Packaging | Ampoule |
| Form | Liquid |