missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc, Zn/Al/Cu, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Zinc, Zn/Al/Cu, Ø50x20mm
Specifications
Specifications
| Chemical Name or Material | Manganese/Chromium/Nickel/Cadmium/Magnesium/Lead/Copper/Aluminium/Tin/Iron |
| CAS | 7440-66-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Manganese 0.00021 %/Chromium 0.00024 %/Nickel 0.00033 %/Cadmium 0.0009 %/Magnesium 0.00155 %/Lead 0.0027 %/Copper 0.481 %/Aluminium 2.51 %/Tin (0.0005) %/Iron (0.0017) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction