missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMYM4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94676-0.1ml
This item is not returnable.
View return policy
Description
ZMYM4 Polyclonal antibody specifically detects ZMYM4 in Human samples. It is validated for Western Blot
Specifications
| ZMYM4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CDIR, cell death inhibiting RNA, KIAA0425DKFZp686L0547, MYM, zinc finger MYM-type protein 4, Zinc finger protein 262DKFZp686B09210, zinc finger, MYM-type 4, ZNF198L3, ZNF262 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 35-230 of human ZMYM4 (NP_005086.2). TEMSEDIDHNLTPTLDSMSYGMPNQTGSENSLLDEDDYFLNSGDLAGIPVVGSDNEDEQDFSSKDNLVSSIHTDDSLEVERRVTQHESDNENEIQIQNKLKKDFPKQFDQVSVFKSIRKDFSLVRENSKETFSGKEKNRDLTYEREKRLDKPHKDLDSRLKSSFFDKAANQVEETLHTHLPQTPETNFRDSSYPFA | |
| 0.1 mL | |
| Cell Biology | |
| 9202 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction