missing translation for 'onlineSavingsMsg'
Learn More

ZMYM4 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 198.00 - € 468.00

Specifications

Antigen ZMYM4
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18673391
View Documents
Novus Biologicals
NBP2-94676-0.02ml
0.02 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18619970
View Documents
Novus Biologicals
NBP2-94676-0.1ml
0.1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

ZMYM4 Polyclonal antibody specifically detects ZMYM4 in Human samples. It is validated for Western Blot
Specifications

Specifications

ZMYM4
Western Blot
Unconjugated
Rabbit
Cell Biology
PBS (pH 7.3), 50% glycerol
9202
IgG
Affinity purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human
CDIR, cell death inhibiting RNA, KIAA0425DKFZp686L0547, MYM, zinc finger MYM-type protein 4, Zinc finger protein 262DKFZp686B09210, zinc finger, MYM-type 4, ZNF198L3, ZNF262
Recombinant fusion protein containing a sequence corresponding to amino acids 35-230 of human ZMYM4 (NP_005086.2). TEMSEDIDHNLTPTLDSMSYGMPNQTGSENSLLDEDDYFLNSGDLAGIPVVGSDNEDEQDFSSKDNLVSSIHTDDSLEVERRVTQHESDNENEIQIQNKLKKDFPKQFDQVSVFKSIRKDFSLVRENSKETFSGKEKNRDLTYEREKRLDKPHKDLDSRLKSSFFDKAANQVEETLHTHLPQTPETNFRDSSYPFA
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.