Learn More
ZNFX1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55381-1-AP-150UL
 View more versions of this product
Product Code. 18030476
  • € 410.00 / 150µL
Description
The ZNFX1 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human ZNFX1. This antibody recognizes human antigen. The ZNFX1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ZNFX1
1000 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
KIAA1404, ZNFX1
150 μL
57169
Store at -20°C. Stable for one year after shipment.