Learn More
ZNFX1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | ZNFX1 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18038175
|
Proteintech
55381-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18030476
|
Proteintech
55381-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ZNFX1 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human ZNFX1. This antibody recognizes human antigen. The ZNFX1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ZNFX1 | |
| Western Blot, ELISA | |
| Rabbit | |
| 57169 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 1000 μg/mL | |
| Liquid | |
| Human | |
| KIAA1404, ZNFX1 | |
| Primary |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.