missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNRF1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 468.00
Specifications
| Antigen | ZNRF1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ZNRF1 Polyclonal antibody specifically detects ZNRF1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ZNRF1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| PBS (pH 7.3), 50% glycerol | |
| 84937 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp434E229, EC 6.3.2, EC 6.3.2.-, FLJ14846, MGC15430, nerve injury gene 283, Nerve injury-induced gene 283 protein, nin283, NIN283E3 ubiquitin-protein ligase ZNRF1, zinc and ring finger 1, zinc and ring finger protein 1, Zinc/RING finger protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human ZNRF1 (NP_115644.1). MGGKQSTAARSRGPFPGVSTDDSAVPPPGGAPHFGHYRTGGGAMGLRSRSVSSVAGMGMDPSTAGGVPFGLYTPASRGTGDSERAPGGGGSASDSTYAHGNGYQETGGGHHRDGMLYLGSRASLADALPLHIAPRWFSSHSGFKCPICSKSVASDEMEMHFIMCLSKPRL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title