missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZSCAN1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94744-0.02ml
This item is not returnable.
View return policy
Description
ZSCAN1 Polyclonal antibody specifically detects ZSCAN1 in Human samples. It is validated for Western Blot
Specifications
| ZSCAN1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ33779, MGC104472, MZF-1, zinc finger and SCAN domain containing 1, zinc finger and SCAN domain-containing protein 1, zinc finger with SCAN domain 1, ZNF915 | |
| A synthetic peptide corresponding to a sequence within amino acids 300-400 of human ZSCAN1 (NP_872378.3). VFTWVTHFIEHQKTHREEGPFPCPECGKVFLHNSVLTEHGKIHLLEPPRKKAPRSKGPRESVPPRDGAQGPVAPRSPKRPFQCSVCGKAFPWMVHLIDHQK | |
| 0.02 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 284312 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction