missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZSCAN1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 221.00 - € 470.00
Specifications
| Antigen | ZSCAN1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18682252
|
Novus Biologicals
NBP2-94744-0.02ml |
0.02 mL |
€ 221.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18687091
|
Novus Biologicals
NBP2-94744-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZSCAN1 Polyclonal antibody specifically detects ZSCAN1 in Human samples. It is validated for Western BlotSpecifications
| ZSCAN1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| FLJ33779, MGC104472, MZF-1, zinc finger and SCAN domain containing 1, zinc finger and SCAN domain-containing protein 1, zinc finger with SCAN domain 1, ZNF915 | |
| A synthetic peptide corresponding to a sequence within amino acids 300-400 of human ZSCAN1 (NP_872378.3). VFTWVTHFIEHQKTHREEGPFPCPECGKVFLHNSVLTEHGKIHLLEPPRKKAPRSKGPRESVPPRDGAQGPVAPRSPKRPFQCSVCGKAFPWMVHLIDHQK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 284312 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.